Learn More
MSC Quantum Storage Plastic Drawer Bin, High-Impact Polystyrene, Yellow, 5-9/16 in Width
Catalog No. 5000511393
Encompass
This plastic drawer bin is constructed from durable high-impact polystyrene, offering waterproof and chemical-resistant properties. Its yellow color makes it easily identifiable, and it features a smooth front curved pull handle for convenient use. Designed as a rectangular drawer-style bin, it is suitable for various organizational needs.
  • Waterproof design
  • Full width back ledge
  • Front handle has 3/4 in opening
  • Oil and chemical resistant
  • Smooth front curved pull handle
  • For use with open and closed shelf units and pick racks
Catalog No. 50-005-11393 Supplier MSC Supplier No. 89826309
This plastic drawer bin is constructed from durable high-impact polystyrene, offering waterproof and chemical-resistant properties. Its yellow color makes it easily identifiable, and it features a smooth front curved pull handle for convenient use. Designed as a rectangular drawer-style bin, it is suitable for various organizational needs.
  • Waterproof design
  • Full width back ledge
  • Front handle has 3/4 in opening
  • Oil and chemical resistant
  • Smooth front curved pull handle
  • For use with open and closed shelf units and pick racks