Learn More
MSC Quantum Storage Plastic Hopper Shelf Bin, Polypropylene, Red, 50 Lb Capacity
Catalog No. 5000511952
Encompass
This hopper shelf bin is constructed from durable polypropylene plastic, offering a 50 Lb capacity. It is designed for efficient storage and organization, featuring reinforced edges and a built-in rear hang-lock for secure placement.
  • Molded-in label holder
  • Reinforced edges
  • Tough, durable, high density economy construction
  • Built-in rear hang-lock
  • 50 Lb capacity
  • Suitable for use with 12 in, 18 in, and 24 in deep shelf units, racks, or standard shelving
Catalog No. 50-005-11952 Supplier MSC Supplier No. 73087322
This hopper shelf bin is constructed from durable polypropylene plastic, offering a 50 Lb capacity. It is designed for efficient storage and organization, featuring reinforced edges and a built-in rear hang-lock for secure placement.
  • Molded-in label holder
  • Reinforced edges
  • Tough, durable, high density economy construction
  • Built-in rear hang-lock
  • 50 Lb capacity
  • Suitable for use with 12 in, 18 in, and 24 in deep shelf units, racks, or standard shelving