Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Polyethylene, Red, 75 lb
Catalog No. 5000514355
Encompass
This plastic hopper stacking bin from Quantum Storage is designed for efficient organization and storage in various settings. Made from durable polyethylene, it offers a 75 lb capacity and is resistant to rust and corrosion, ensuring longevity and ease of maintenance. Its stackable design and integrated features make it a versatile storage solution.
  • Hopper stacking bin style
  • Rectangle shape
  • Large label holder
  • Easy to clean
  • Resistant to rust and corrosion
  • Six 1/4 in drill hole pattern on bottom
  • Stacking capable
  • Compatible with optional window accessories
  • Designed for use with tall and sturdy storage systems
Catalog No. 50-005-14355 Supplier MSC Supplier No. 86550241
This plastic hopper stacking bin from Quantum Storage is designed for efficient organization and storage in various settings. Made from durable polyethylene, it offers a 75 lb capacity and is resistant to rust and corrosion, ensuring longevity and ease of maintenance. Its stackable design and integrated features make it a versatile storage solution.
  • Hopper stacking bin style
  • Rectangle shape
  • Large label holder
  • Easy to clean
  • Resistant to rust and corrosion
  • Six 1/4 in drill hole pattern on bottom
  • Stacking capable
  • Compatible with optional window accessories
  • Designed for use with tall and sturdy storage systems