Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Polypropylene, Clear, 75 Lb
Catalog No. 5000535279
Encompass
This plastic hopper stacking bin is made from clear polypropylene with a 75 Lb capacity, designed for efficient storage and organization.
  • Clear styrene window
  • Front, back & side grip
  • Waterproof bins
  • Resistant to extreme cold
  • Resistant rust & corrosion
  • Autoclavable up to 250 °F
  • Molded ID card slot
Catalog No. 50-005-35279 Supplier MSC Supplier No. 07295512
This plastic hopper stacking bin is made from clear polypropylene with a 75 Lb capacity, designed for efficient storage and organization.
  • Clear styrene window
  • Front, back & side grip
  • Waterproof bins
  • Resistant to extreme cold
  • Resistant rust & corrosion
  • Autoclavable up to 250 °F
  • Molded ID card slot