Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Polypropylene, Clear, Autoclavable
Catalog No. 5000510761
Encompass
This plastic hopper stacking bin is constructed from polypropylene and features a clear design. It is suitable for use with louvered panels or rails, offering both stacking and hanging capabilities. The bin has overall dimensions of 14-3/4 inches in length, 6-9/16 inches in width, and 7 inches in height, with corresponding inside dimensions of 14 inches in length, 6-9/16 inches in width, and 6-3/4 inches in height.
  • Clear styrene window for easy visibility of contents.
  • Front, back, and side grip for comfortable handling.
  • Waterproof construction.
  • Resistant to extreme cold, rust, and corrosion.
  • Autoclavable up to 250°F.
  • Molded ID card slot for content identification.
  • Designed for use with louvered panels or rails.
Catalog No. 50-005-10761 Supplier MSC Supplier No. 58662800
This plastic hopper stacking bin is constructed from polypropylene and features a clear design. It is suitable for use with louvered panels or rails, offering both stacking and hanging capabilities. The bin has overall dimensions of 14-3/4 inches in length, 6-9/16 inches in width, and 7 inches in height, with corresponding inside dimensions of 14 inches in length, 6-9/16 inches in width, and 6-3/4 inches in height.
  • Clear styrene window for easy visibility of contents.
  • Front, back, and side grip for comfortable handling.
  • Waterproof construction.
  • Resistant to extreme cold, rust, and corrosion.
  • Autoclavable up to 250°F.
  • Molded ID card slot for content identification.
  • Designed for use with louvered panels or rails.