Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Polypropylene, Yellow, 100 lb Capacity
Catalog No. 5000527976
Encompass
Quantum Storage plastic hopper stacking bins are constructed from durable polypropylene, offering a robust solution for storage and organization. These yellow bins are designed to be stackable and are resistant to various environmental conditions, including extreme cold and corrosion. They feature a 100 lb capacity, making them suitable for demanding laboratory or industrial applications.
  • Heavy-duty front, back, and side grip
  • Clear plastic window
  • Waterproof and corrosion-free
  • Resistant to extreme cold
  • Stackable design
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F
Catalog No. 50-005-27976 Supplier MSC Supplier No. 73087660
Quantum Storage plastic hopper stacking bins are constructed from durable polypropylene, offering a robust solution for storage and organization. These yellow bins are designed to be stackable and are resistant to various environmental conditions, including extreme cold and corrosion. They feature a 100 lb capacity, making them suitable for demanding laboratory or industrial applications.
  • Heavy-duty front, back, and side grip
  • Clear plastic window
  • Waterproof and corrosion-free
  • Resistant to extreme cold
  • Stackable design
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F