Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Polypropylene, Yellow, 30 Lb Capacity
Catalog No. 5000534758
Encompass
Quantum Storage Plastic Hopper Stacking Bins are designed for efficient organization and storage. Made from durable polypropylene, these yellow bins are ideal for various industrial and laboratory settings. They feature a hopper design for easy access to contents, a large molded front label area for clear identification, and are resistant to water, rust, and corrosion.
  • Crystal styrene window
  • FDA approved
  • Water, rust, and corrosion proof
  • Front, back, and side grip
  • Large molded front label
  • Resistant to extreme cold
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F
  • Stackable design
  • Hanging capability
Catalog No. 50-005-34758 Supplier MSC Supplier No. 73087975
Quantum Storage Plastic Hopper Stacking Bins are designed for efficient organization and storage. Made from durable polypropylene, these yellow bins are ideal for various industrial and laboratory settings. They feature a hopper design for easy access to contents, a large molded front label area for clear identification, and are resistant to water, rust, and corrosion.
  • Crystal styrene window
  • FDA approved
  • Water, rust, and corrosion proof
  • Front, back, and side grip
  • Large molded front label
  • Resistant to extreme cold
  • Wide stacking ledge and anti-slide lock
  • Autoclavable up to 250°F
  • Stackable design
  • Hanging capability