Learn More
MSC Quantum Storage Plastic Hopper Stacking Bin, Red, Polyethylene, 100 Lb Capacity
Catalog No. 5000534620
Encompass
This plastic hopper stacking bin is designed for efficient organization and storage of various items. Constructed from heavy-duty polyethylene, it offers excellent durability and resistance to solvents, corrosion, rust, and rot. The bin features a rear molded handle for easy handling and can withstand temperatures ranging from -40°F to 250°F. Ideal for use with wire shelving and steel shelving systems, this bin helps maintain an organized workspace.
  • Heavy-duty, extra strong injection molded construction.
  • Resistant to solvents, corrosion, rust, and rot.
  • Withstands temperatures from -40°F to 250°F.
  • Features a rear molded handle for easy transport.
  • Stackable design for space-saving storage.
Catalog No. 50-005-34620 Supplier MSC Supplier No. 73087116
This plastic hopper stacking bin is designed for efficient organization and storage of various items. Constructed from heavy-duty polyethylene, it offers excellent durability and resistance to solvents, corrosion, rust, and rot. The bin features a rear molded handle for easy handling and can withstand temperatures ranging from -40°F to 250°F. Ideal for use with wire shelving and steel shelving systems, this bin helps maintain an organized workspace.
  • Heavy-duty, extra strong injection molded construction.
  • Resistant to solvents, corrosion, rust, and rot.
  • Withstands temperatures from -40°F to 250°F.
  • Features a rear molded handle for easy transport.
  • Stackable design for space-saving storage.