Learn More
MSC Quantum Storage Polyethylene Storage Tote, Polyethylene, 100 Lb Capacity, Stacking, Nesting
Catalog No. 5000514211
Encompass
This storage tote is made with injection molded, high-density polyethylene that withstands temperatures from -10°F to 250°F. It complies with USDA and FDA regulations for use in food processing applications. The plastic tote won't crack or corrode and is waterproof to ensure long life. It features 180 degree nesting and stacking to save valuable storage space, fits on standard 48" x 40" pallets, and has a textured bottom for a non-slip grip during transport and on conveyors.
  • Reusable container
  • Suitable for storage and transportation of goods
  • Secures valuable items
  • Available in multiple colors for inventory control
  • Optional lid snaps keep stored items secure
  • Designed for stacking with other containers to save space
  • Nestable when not in use to save space
Catalog No. 50-005-14211 Supplier MSC Supplier No. 86550480
This storage tote is made with injection molded, high-density polyethylene that withstands temperatures from -10°F to 250°F. It complies with USDA and FDA regulations for use in food processing applications. The plastic tote won't crack or corrode and is waterproof to ensure long life. It features 180 degree nesting and stacking to save valuable storage space, fits on standard 48" x 40" pallets, and has a textured bottom for a non-slip grip during transport and on conveyors.
  • Reusable container
  • Suitable for storage and transportation of goods
  • Secures valuable items
  • Available in multiple colors for inventory control
  • Optional lid snaps keep stored items secure
  • Designed for stacking with other containers to save space
  • Nestable when not in use to save space