Learn More
MSC Quantum Storage Solid Shelf, Stainless Steel, 36 in, 14 in
Catalog No. 5000512307
Encompass
This solid stainless steel shelf is constructed from durable 304 stainless steel, offering excellent corrosion resistance and easy cleaning. Measuring 36 inches wide by 14 inches deep, it is designed for use in demanding environments such as professional kitchens and storage rooms. The shelf features a double-hemmed perimeter with a marine edge, effectively containing spills.
  • Made from durable 304 stainless steel
  • Features a double-hemmed perimeter with a marine edge
  • Easy to clean
  • Certified by NSF International to meet applicable sanitation and food safety standards
  • Withstands high moisture and cold weather environments
  • Has a load capacity of 600 lb
Catalog No. 50-005-12307 Supplier MSC Supplier No. 91530337
This solid stainless steel shelf is constructed from durable 304 stainless steel, offering excellent corrosion resistance and easy cleaning. Measuring 36 inches wide by 14 inches deep, it is designed for use in demanding environments such as professional kitchens and storage rooms. The shelf features a double-hemmed perimeter with a marine edge, effectively containing spills.
  • Made from durable 304 stainless steel
  • Features a double-hemmed perimeter with a marine edge
  • Easy to clean
  • Certified by NSF International to meet applicable sanitation and food safety standards
  • Withstands high moisture and cold weather environments
  • Has a load capacity of 600 lb