This storage tote is constructed from injection molded, high-density polypropylene, withstanding temperatures from -10°F to 250°F. It meets USDA and FDA food processing regulations, offering waterproof protection and a long service life. Its 180-degree nest and stack capabilities optimize space, and a textured bottom provides a non-slip grip during transport.
- Reusable container for versatile applications.
- Secure storage and transport for goods.
- Available in gray, blue, and red for inventory management.
- Optional lid snaps ensure item security.
- Stackable and nestable design saves space.
- Withstands temperatures from -10°F to 250°F.
- USDA and FDA compliant for food processing.
- Waterproof, preventing cracking or corrosion.
- Textured bottom for non-slip handling.
- Fits standard 48 in x 40 in pallets.
- 75 lb load capacity.