Learn More
MSC Quantum Storage Systems Floor Pick Rack, 2100 Lb Capacity, 48 Bins, Steel, Ivory
Catalog No. 5000535610
Encompass
This floor pick rack provides efficient storage and organization for various items. Constructed with a durable steel frame, it is designed for demanding environments and offers a substantial 2,100 Lb load capacity. The rack includes 48 ivory bins made from a robust polyethylene and polypropylene blend, ideal for sorting and accessing components easily. Its compact dimensions make it suitable for a variety of floor layouts.
  • 2,100 Lb load capacity
  • 12 in deep x 36 in wide x 39 in high dimensions
  • Steel rack material
  • Polyethylene and polypropylene bin material
  • 48 bins included
  • Ivory bin color
  • Single-sided design
Catalog No. 50-005-35610 Supplier MSC Supplier No. 48519987
This floor pick rack provides efficient storage and organization for various items. Constructed with a durable steel frame, it is designed for demanding environments and offers a substantial 2,100 Lb load capacity. The rack includes 48 ivory bins made from a robust polyethylene and polypropylene blend, ideal for sorting and accessing components easily. Its compact dimensions make it suitable for a variety of floor layouts.
  • 2,100 Lb load capacity
  • 12 in deep x 36 in wide x 39 in high dimensions
  • Steel rack material
  • Polyethylene and polypropylene bin material
  • 48 bins included
  • Ivory bin color
  • Single-sided design