Learn More
MSC Quantum Storage Systems Snap on Cover, Clear, Polypropylene, 11 in L x 8 in W x 1.05 in H
Catalog No. 5000531122
Encompass
The Snap on Cover is designed for use with dividable grid containers. It provides protection for the interior of bins from dust, dirt, and moisture, ensuring contents remain clean. Made from durable polypropylene, this clear cover can be easily washed with water and normal cleaners.
  • Made of polypropylene
  • Clear color
  • Protects interior of bins from dust, dirt, and moisture
  • Can be washed with water and normal cleaners
  • Designed for dividable grid containers
Catalog No. 50-005-31122 Supplier MSC Supplier No. 86550597
The Snap on Cover is designed for use with dividable grid containers. It provides protection for the interior of bins from dust, dirt, and moisture, ensuring contents remain clean. Made from durable polypropylene, this clear cover can be easily washed with water and normal cleaners.
  • Made of polypropylene
  • Clear color
  • Protects interior of bins from dust, dirt, and moisture
  • Can be washed with water and normal cleaners
  • Designed for dividable grid containers