Learn More
MSC Quantum Storage Systems Ultra Stack and Hang Bin, Polypropylene, 18 x 8.25 x 9 in, Ivory
Catalog No. 5000534740
Encompass
Quantum Storage Systems Ultra Stack and Hang Bins are heavy-duty storage containers designed for organizing work and storage areas. Constructed from durable polypropylene, these bins are suitable for various industrial and laboratory applications, offering resistance to chemicals and extreme temperatures.
  • Autoclavable up to 250°F and resistant to extreme cold.
  • Easily cleaned with water and soap.
  • Impervious to most chemicals, water, rust, and corrosion.
  • Manufactured from FDA approved materials.
  • Stackable design with heavy-duty front, back, and side grips.
  • Features a wide stacking ledge and anti-slide lock.
  • Includes a divider slot, rear hanger, and molded-in label holder.
Catalog No. 50-005-34740 Supplier MSC Supplier No. 73088270
Quantum Storage Systems Ultra Stack and Hang Bins are heavy-duty storage containers designed for organizing work and storage areas. Constructed from durable polypropylene, these bins are suitable for various industrial and laboratory applications, offering resistance to chemicals and extreme temperatures.
  • Autoclavable up to 250°F and resistant to extreme cold.
  • Easily cleaned with water and soap.
  • Impervious to most chemicals, water, rust, and corrosion.
  • Manufactured from FDA approved materials.
  • Stackable design with heavy-duty front, back, and side grips.
  • Features a wide stacking ledge and anti-slide lock.
  • Includes a divider slot, rear hanger, and molded-in label holder.