Learn More
MSC Quantum Storage Wire Basket Wire Shelving, Chrome Wire, 60 x 18 x 10 in, 800 Lb Load Capacity
Catalog No. 5000536447
Encompass
This Quantum Storage Wire Basket Wire Shelving unit is designed for efficient storage. It features durable chrome wire construction and offers a substantial 800 Lb load capacity. With dimensions of 60 in wide, 18 in deep, and 10 in high, it is suitable for various storage applications.
  • Add-on shelving component
  • Measures 60 in wide, 18 in deep, and 10 in high
  • 800 Lb load capacity
  • Durable chrome wire construction
  • Country of origin: China
Catalog No. 50-005-36447 Supplier MSC Supplier No. 91530766
This Quantum Storage Wire Basket Wire Shelving unit is designed for efficient storage. It features durable chrome wire construction and offers a substantial 800 Lb load capacity. With dimensions of 60 in wide, 18 in deep, and 10 in high, it is suitable for various storage applications.
  • Add-on shelving component
  • Measures 60 in wide, 18 in deep, and 10 in high
  • 800 Lb load capacity
  • Durable chrome wire construction
  • Country of origin: China