Learn More
BANGS LABORATORIES INC Quantum QC, 8 Peak Beads, 5 mL
Catalog No. NC0898256
Encompass
Quantum QC is a 8 peak multi-intensity, multi-fluorescent standard that is intended for use as an in depth tool for daily cytometer QC, and is appropriate for use with all lasers and detectors.
Catalog No. NC0898256 Supplier BANGS LABORATORIES INC Supplier No. 725
Quantum QC is a 8 peak multi-intensity, multi-fluorescent standard that is intended for use as an in depth tool for daily cytometer QC, and is appropriate for use with all lasers and detectors.