Learn More
Andwin Scientific QUARTER RESERVOIR 40ML 48PK
SDP
Catalog No. NC9969411
Encompass_Preferred
The Biomek Modular Quarter Reservoir can hold liquid for pipetting using a manual pipettor or liquid handling system.This product requires a frame that is able to accommodate up to four quarter reservoirs. The reservoir has a volume capacity of 40 mL and is sold in a case of 48.
Catalog No. NC9969411 Supplier Andwin Scientific Supplier No. BK372790
The Biomek Modular Quarter Reservoir can hold liquid for pipetting using a manual pipettor or liquid handling system.This product requires a frame that is able to accommodate up to four quarter reservoirs. The reservoir has a volume capacity of 40 mL and is sold in a case of 48.