Learn More
Enterprise Technology Solutions Quartet Presentation Easel, Whiteboard/Flipchart, 3'x2', WE/GY
SDP
Supplier:  Enterprise Technology Solutions QRTET32EU
Encompass_Preferred
Standing nearly 75" tall at full height, this presentation easel features a smooth, melamine whiteboard writing surface that's durable enough for light use and provides a sturdy backing for flipchart usage. Telescopic legs with rubber feet are individually height-adjustable to ensure your message can be seen from anywhere in the room. Full-width accessory tray provides quick access to your dry-erase markers and accessories. Collapsible design allows easy mobility and compact storage. Presentation easel with flipchart holder is perfect for presentations, collaborating with your team, brainstorming and more.
Catalog No. 50-007-90988