Learn More
Enterprise Technology Solutions Quartet Tripod Easel, Steel, Foldable, 72", Bronze
SDP
Catalog No. 5000791928
Encompass_Preferred
Tripod easel is made of sturdy steel with bronze finish. Locking mechanism secures legs. Included two sets of brackets fold down to hold presentation materials. Tripod easel folds to 1" wide x 4" deep x 72" high for easy transport or storage. Easel supports displays up to 8' x 4'.
Catalog No. 50-007-91928 Supplier Enterprise Technology Solutions Supplier No. QRT21E
Tripod easel is made of sturdy steel with bronze finish. Locking mechanism secures legs. Included two sets of brackets fold down to hold presentation materials. Tripod easel folds to 1" wide x 4" deep x 72" high for easy transport or storage. Easel supports displays up to 8' x 4'.