Learn More
Perkin Elmer US LLC Quartz 10 mm Bending Platform for DMA 7/DMA 7e 3-Point Bending Measuring System and TMA 7
Catalog No. 502113568
Encompass_Preferred
This tube differs from the standard sample tube in that the 10mm bending platform is permanently fused to the base of the sample tube. Used with the high temperature furnace.
Catalog No. 50-211-3568 Supplier Perkin Elmer US LLC Supplier No. N5391181
This tube differs from the standard sample tube in that the 10mm bending platform is permanently fused to the base of the sample tube. Used with the high temperature furnace.