Learn More
Sigma Aldrich Fine Chemicals Biosciences Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 2 mm, chamber volume 700 muL | 1EA
Catalog No. NC1682938
Encompass
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 2 mm, chamber volume 700 muL | 1EA
Catalog No. NC1682938 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8001201EA
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2500 nm, pathlength 2 mm, chamber volume 700 muL | 1EA