Learn More
Perkin Elmer US LLC Quartz Baffled Cyclonic Spray Chamber without Gas Port for ELAN 5000/6x00/9000/DRCs PC3 Systems
Catalog No. 502113241
Encompass_Preferred
The quartz baffled cyclonic spray chamber features a 7 mm baffle and auxiliary gas port. It's designed for all-purpose, non-hydrofluoric acid resistant applications and provides excellent sensitivity and stability with low background.
Catalog No. 50-211-3241 Supplier Perkin Elmer US LLC Supplier No. N0777035
The quartz baffled cyclonic spray chamber features a 7 mm baffle and auxiliary gas port. It's designed for all-purpose, non-hydrofluoric acid resistant applications and provides excellent sensitivity and stability with low background.