Learn More
Perkin Elmer US LLC Quartz Torch Bonnet for Optima 2x00/4x00/5x00/7x00 DV
Catalog No. 509050156
Encompass_Preferred
The torch bonnet is used as a spacer between the torch and RF load coil. This bonnet is included in the Organics Kit for Optima 4300 V/5300 V/7300 V(N0770732), the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770731), and the Cyclonic/Concentric Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770730).
Catalog No. 50-905-0156 Supplier Perkin Elmer US LLC Supplier No. N0775289
The torch bonnet is used as a spacer between the torch and RF load coil. This bonnet is included in the Organics Kit for Optima 4300 V/5300 V/7300 V(N0770732), the Scott/Cross-Flow II Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770731), and the Cyclonic/Concentric Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770730).