Learn More
Glass Expansion Capillary Quartz Injector 3.0mm ID
SDP
Catalog No. 502703140
Encompass_Preferred
Capillary Quartz Injector 3.0mm ID. Compatible with Glass Expansion’s D-Torch and EMT Torch for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES. Suitable for use with Glass Expansion D-Torches NC2046971, 30-808-2877, 501694650. In general, quartz injectors are designed for organic solvents and non-HF matrices. A 3.0mm large bore injector is best suited for aqueous and non-HF matrices with high TDS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3140 Supplier Glass Expansion Supplier No. 318083247
Capillary Quartz Injector 3.0mm ID. Compatible with Glass Expansion’s D-Torch and EMT Torch for Thermo PRO Duo, PRO Radial, 6000/7000 Duo & Radial Series ICP-OES. Suitable for use with Glass Expansion D-Torches NC2046971, 30-808-2877, 501694650. In general, quartz injectors are designed for organic solvents and non-HF matrices. A 3.0mm large bore injector is best suited for aqueous and non-HF matrices with high TDS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001