Learn More
Thermo Scientific Quartz center tube
Catalog No. NC1977476 Shop All Thermo Scientific Products
Encompass
Quartz center tube has 2.0 mm outer diameter, provides high heat resistance, chemical inertness, optical transparency, with inner bore passing gases, liquids, light for analytical measurements, used as sample chamber, waveguide in spectrometers, heaters, laboratory equipment.
Catalog No. NC1977476 Supplier Thermo Scientific Supplier No. BRE0019895
Quartz center tube has 2.0 mm outer diameter, provides high heat resistance, chemical inertness, optical transparency, with inner bore passing gases, liquids, light for analytical measurements, used as sample chamber, waveguide in spectrometers, heaters, laboratory equipment.