Learn More
Anton Paar QUARTZ CONTROL PLATE, +50°Z/+17°OR for MCP 150
Catalog No. NC1779946
Encompass_Preferred
Quartz control plate with +50°Z/+17°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 150.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945
This item is not returnable. View return policy
Catalog No. NC1779946 Supplier Anton Paar Supplier No. 104461
Quartz control plate with +50°Z/+17°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 150.Suitable for use with Anton Paar polarimeter MCP 150 item NC1779945