Learn More
Anton Paar QUARTZ CONTROL PLATE, +100°Z/+17°OR for MCP 100
Catalog No. NC1818317
Encompass_Preferred
Quartz control plate with +100°Z/+34°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 100.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306
Catalog No. NC1818317 Supplier Anton Paar Supplier No. 143961
Quartz control plate with +100°Z/+34°Optical Rotation used for checking and adjusting the polarimeter scale on the MCP 100.Suitable for use with Anton Paar polarimeter MCP 100 item NC1818306