Learn More
Polysciences, Inc. Quartz Coverslip,1.0" (25.4 mm) Square, Thickness #1 (0.15 to 0.18 mm) SOLD IND.
SDP
Catalog No. NC1220584
Encompass_Preferred
Quartz Coverslip,1.0" (25.4 mm) Square, Thickness #1 (0.15 to 0.18 mm) SOLD IND.
Catalog No. NC1220584 Supplier Polysciences, Inc. Supplier No. 249681
Quartz Coverslip,1.0" (25.4 mm) Square, Thickness #1 (0.15 to 0.18 mm) SOLD IND.