Learn More
Chemglass Life Sciences 19mm X 19mm Quartz Cover Slip
SDP
Catalog No. CGQ066001
Encompass_Preferred
19mm X 19mm Quartz Cover Slip. Ultra-thin (.25mm) ground and polished clear quartz microscope slide cover slips.
Catalog No. CGQ066001 Supplier Chemglass Life Sciences Supplier No. CGQ066001
19mm X 19mm Quartz Cover Slip. Ultra-thin (.25mm) ground and polished clear quartz microscope slide cover slips.