Learn More
Electron Microscopy Sciences Quartz Coverslip Square #2, 25 mm
SDP
Catalog No. 501927082
Encompass_Preferred
There are many applications that require slides and coverslips that have absolute UV transparency, and high temperature properties. EMS now is introducing the highest quality, clear fused quartz microscope slides and coverslips to meet all of these requirements. Fused quartz microscope slides and coverslips with a window of highest transparency to UV is between 0.15µ to 5µ. Typical flatness measured 2 to 4 bands per inch and soften point is about 1250 degree C.
Size: 25 mm
Thickness: 0.2 mm
Catalog No. 50-192-7082 Supplier Electron Microscopy Sciences Supplier No. 7225607
There are many applications that require slides and coverslips that have absolute UV transparency, and high temperature properties. EMS now is introducing the highest quality, clear fused quartz microscope slides and coverslips to meet all of these requirements. Fused quartz microscope slides and coverslips with a window of highest transparency to UV is between 0.15µ to 5µ. Typical flatness measured 2 to 4 bands per inch and soften point is about 1250 degree C.
Size: 25 mm
Thickness: 0.2 mm