Learn More
CEM Corporation QUARTZ CRUCIBLE 50/CS
SDP
Catalog No. NC0652009
Encompass
CEM's unique, patented quartz fiber crucibles reduce ashing times by as much as 97% and cool in seconds. The quartz fiber material allows oxygen to circulate around the sample speeding combustion even with our unique crucible caps in place. These unbreakable crucibles withstand temperatures up to 1,000 deg.C and do not need desiccation. Quartz fiber crucible caps can be used with our 50- or 100-ml crucibles. Try them out next time you have to ash a sample that needs to be contained. Ideal for low ash samples and samples with high organic content, they reduce the chance for sample contamination and improve accuracy on ROI and LOI determinations.
Catalog No. NC0652009 Supplier CEM Corporation Supplier No. 303060
CEM's unique, patented quartz fiber crucibles reduce ashing times by as much as 97% and cool in seconds. The quartz fiber material allows oxygen to circulate around the sample speeding combustion even with our unique crucible caps in place. These unbreakable crucibles withstand temperatures up to 1,000 deg.C and do not need desiccation. Quartz fiber crucible caps can be used with our 50- or 100-ml crucibles. Try them out next time you have to ash a sample that needs to be contained. Ideal for low ash samples and samples with high organic content, they reduce the chance for sample contamination and improve accuracy on ROI and LOI determinations.