Learn More
Cole-Parmer DWK Life Sciences (Kimble) Quartz Crucible and Cover, 50 mL, 49 mm OD X 50 mm H
Supplier:  Cole-Parmer 3452650
Encompass_Preferred
Quartz crucible satisfies high-temperature requirements up to 1922°F (1050°C)—perfect for muffle furnace ashing procedures. Quartz material ensures chemical purity for subsequent analysis. Crucible is useful for determining trace metals in environmental samples.
Catalog No. NC9318776