Learn More
Technical Glass Products Inc Clear Fused Quartz Crucible w/ Lid 100ML Capcity
SDP
Catalog No. NC0279015
Encompass
Clear Fused Quartz Crucible w/ Lid 100cc capacity, 62mm Top Dia x 66mm Tall, 35mm base diameter
Catalog No. NC0279015 Supplier Technical Glass Products Inc Supplier No. CL 100
Clear Fused Quartz Crucible w/ Lid 100cc capacity, 62mm Top Dia x 66mm Tall, 35mm base diameter