Learn More
Azzota Corp Quartz cuvette 10mm 17.5ml 2wi
Catalog No. 5000887023
Encompass_Preferred
10mm Pathlength Quartz Cuvettes 17 50 ml Dimensions - 45x12 5x52 5mm 190nm-2500nm two large windows clear two small windows frosted
Catalog No. 50-008-87023 Supplier Azzota Corp Supplier No. Q108C
10mm Pathlength Quartz Cuvettes 17 50 ml Dimensions - 45x12 5x52 5mm 190nm-2500nm two large windows clear two small windows frosted