Learn More
Cole-Parmer Jenway Quartz Cuvette, Semi-Micro, UV/Visible, 1 mL, 10 mm Path; 1/EA
Catalog No. NC1485831 Shop All Cole-Parmer Products
Encompass_Preferred
Jenway Quartz Semi-micro Cuvette, 1 mL, 10 mm path, Suitable for both UV and visible regions, Quartz cuvettes are available in a wide capacity range from 50 L to 35 mL and path lengths from 10 to 100 mm. Visible % transmission: maximum from 400 to 750 nmUV wavelength % transmission: maximum from 200 to 400 nm
Catalog No. NC1485831 Supplier Cole-Parmer Supplier No. 8307086
Jenway Quartz Semi-micro Cuvette, 1 mL, 10 mm path, Suitable for both UV and visible regions, Quartz cuvettes are available in a wide capacity range from 50 L to 35 mL and path lengths from 10 to 100 mm. Visible % transmission: maximum from 400 to 750 nmUV wavelength % transmission: maximum from 200 to 400 nm