Learn More
Azzota Corp Quartz cuvette 20mm 7ml 190-25
Catalog No. 5000887059
Encompass_Preferred
20mm Pathlength Quartz Standard Cuvettes 7 00 ml Dimensions - 45x12 5x22 5mm 190nm-2500nm
Catalog No. 50-008-87059 Supplier Azzota Corp Supplier No. Q105
20mm Pathlength Quartz Standard Cuvettes 7 00 ml Dimensions - 45x12 5x22 5mm 190nm-2500nm