Learn More
Cole-Parmer Jenway Quartz Cuvette, Ultra-Micro, UV/Visible, 50 uL, 10 mm Path; 1/EA
Catalog No. 501961714 Shop All Cole-Parmer Products
Encompass_Preferred
Quartz cuvettes are available in a wide capacity range from 50 L to 35 mL and path lengths from 10 to 100 mm.
Visible % transmission: maximum from 400 to 750 nm
UV wavelength % transmission: maximum from 200 to 400 nm
Catalog No. 50-196-1714 Supplier Cole-Parmer Supplier No. 8307082
Quartz cuvettes are available in a wide capacity range from 50 L to 35 mL and path lengths from 10 to 100 mm.
Visible % transmission: maximum from 400 to 750 nm
UV wavelength % transmission: maximum from 200 to 400 nm