Learn More
Starna Cells Inc QUARTZ CUVETTE 5CM LIGHTPATH
SDP
Catalog No. NC9067279
Encompass_Preferred
Macro/Standard Rectangular, w/lid, Spectrosil quartz, 9.5 W Internal, 52.5L x 12.5W x 45H, 17.500mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally
Catalog No. NC9067279 Supplier Starna Cells Inc Supplier No. 1Q50
Macro/Standard Rectangular, w/lid, Spectrosil quartz, 9.5 W Internal, 52.5L x 12.5W x 45H, 17.500mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally