Learn More
Andwin Scientific QUARTZ CUVETTES 10MM 2/PK
SDP
Catalog No. NC0252948
Encompass_Preferred
The Spectrosil Spectrophotometer Cells have a pathlength of 10 mm. Its fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain restricts any optical or physical distortion of the two optically
Catalog No. NC0252948 Supplier Andwin Scientific Supplier No. 414004069
The Spectrosil Spectrophotometer Cells have a pathlength of 10 mm. Its fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain restricts any optical or physical distortion of the two optically