Learn More
Perkin Elmer US LLC Quartz Hydride Gas-Liquid Separator for hydrideFAST
Catalog No. 502104138
Encompass_Preferred
This quartz hydride gas-liquid separator is used with all hydrideFAST autosampler systems.
Catalog No. 50-210-4138 Supplier Perkin Elmer US LLC Supplier No. N0782032
This quartz hydride gas-liquid separator is used with all hydrideFAST autosampler systems.