Learn More
Perkin Elmer US LLC Quartz Injector, 0.80 mm I.D. for Optima 8x00
Catalog No. 502112275
Encompass_Preferred
This Quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.
Catalog No. 50-211-2275 Supplier Perkin Elmer US LLC Supplier No. N0781018
This Quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.