Learn More
Perkin Elmer US LLC Quartz Injector, 0.80 mm I.D. for Avio
Catalog No. 502113584
Encompass_Preferred
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.
Catalog No. 50-211-3584 Supplier Perkin Elmer US LLC Supplier No. N0791185
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.