Learn More
Perkin Elmer US LLC Quartz Injector, 1.2 mm I.D. - Entire Length of Tube, Optima 8x00
Catalog No. 502113140
Encompass_Preferred
This Quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.
Catalog No. 50-211-3140 Supplier Perkin Elmer US LLC Supplier No. N0781013
This Quartz Injector is for use with Optima 8x00 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.