Learn More
Perkin Elmer US LLC Quartz Injector, 1.6 mm I.D. for Avio
Catalog No. 502113574
Encompass_Preferred
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.
Catalog No. 50-211-3574 Supplier Perkin Elmer US LLC Supplier No. N0791188
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.