Learn More
Glass Expansion Quartz Injector 1.8mm
SDP
Catalog No. 501907216
Encompass_Preferred
Quartz Injector with 1.8mm ID.A 1.8mm ID Quartz Injector (48807023), designed for organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7216 Supplier Glass Expansion Supplier No. 318083391
Quartz Injector with 1.8mm ID.A 1.8mm ID Quartz Injector (48807023), designed for organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001