Learn More
Glass Expansion Quartz Injector 2.0mm
SDP
Catalog No. 501907221
Encompass_Preferred
Quartz Injector with 2.0mm ID.2.0mm ID Quartz Injector (48807024). They are designed for organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7221 Supplier Glass Expansion Supplier No. 318083411
Quartz Injector with 2.0mm ID.2.0mm ID Quartz Injector (48807024). They are designed for organic solvents and non-HF matrices. Compatible with Spectro Blue SOP Torch 50-190-7218 (31-808-3405).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001