Learn More
Glass Expansion Quartz Injector 2.0 mm Thermo Q/RQ/TQ ICP-MS
SDP
Catalog No. 502703112
Encompass_Preferred
Quartz Injector with 2.0mm ID.A 2.0mm ID Quartz Injector (1305640), designed for volatile organic solvents and non-HF matrices. Compatible with Torch Base 501694610 (31-808-3494) for Thermo Q/RQ/TQ.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3112 Supplier Glass Expansion Supplier No. 318083898
Quartz Injector with 2.0mm ID.A 2.0mm ID Quartz Injector (1305640), designed for volatile organic solvents and non-HF matrices. Compatible with Torch Base 501694610 (31-808-3494) for Thermo Q/RQ/TQ.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001