Learn More
Glass Expansion Torch Injector, Glass Expansion, 2.0 mm ID, quartz, for use with D-Torch or single-body quartz torch, EMT (1010501072, 842312051971, BRE0019895)
SDP
Catalog No. 501694654
Encompass_Preferred
Quartz Capillary Injector with 2.0mm ID (842312051971 and BRE0019895).A 2.0mm ID capillary quartz injector. The standard size injector for the Thermo PRO Duo. Suitable for use with aqueous and less-volatile organic solvents-based matrices. Not for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Compatible with Thermo PRO Duo EMT Torch (31-808-4158) and D-Torch (NC2046971 & 30-808-4150), Thermo PRO Radial EMT Torch (50-169-4685 & 31-808-2810) and 6000/7000 Radial D-Torch 30-808-2877, and Thermo 6000/7000 Duo EMT Torch (NC1417608 & 31-808-2811) and D-Torch (50-169-4650 & 30-808-2844).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4654 Supplier Glass Expansion Supplier No. 318082836
Quartz Capillary Injector with 2.0mm ID (842312051971 and BRE0019895).A 2.0mm ID capillary quartz injector. The standard size injector for the Thermo PRO Duo. Suitable for use with aqueous and less-volatile organic solvents-based matrices. Not for use with HF. When replacing your injector, it is recommended to replace the injector ferrule (31-808-2897), on average every 6 months.Compatible with Thermo PRO Duo EMT Torch (31-808-4158) and D-Torch (NC2046971 & 30-808-4150), Thermo PRO Radial EMT Torch (50-169-4685 & 31-808-2810) and 6000/7000 Radial D-Torch 30-808-2877, and Thermo 6000/7000 Duo EMT Torch (NC1417608 & 31-808-2811) and D-Torch (50-169-4650 & 30-808-2844).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001