Learn More
Glass Expansion Torch Injector, Glass Expansion, 2.5 mm ID, quartz
SDP
Catalog No. 501694613
Encompass_Preferred
Quartz Tapered Injector with 2.5mm ID (1305600). A 2.5mm ID quartz tapered injector, ideal for use with non-HF aqueous matrices and high TDS. Not for use with HF. Compatible with Thermo Q/RQ/TQ Semi Demountable Torch (SDT) 50-190-4610 (31-808-3494).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4613 Supplier Glass Expansion Supplier No. 318083428
Quartz Tapered Injector with 2.5mm ID (1305600). A 2.5mm ID quartz tapered injector, ideal for use with non-HF aqueous matrices and high TDS. Not for use with HF. Compatible with Thermo Q/RQ/TQ Semi Demountable Torch (SDT) 50-190-4610 (31-808-3494).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001