Learn More
Perkin Elmer US LLC Quartz Injector, 2.0 mm I.D. for Avio
Catalog No. 502113570
Encompass_Preferred
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.
Catalog No. 50-211-3570 Supplier Perkin Elmer US LLC Supplier No. N0791189
This Quartz Injector is for use with Avio 200 and 500 ICP-OES series instruments. They are designed for organic solvents and non-HF matrices.